Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD63 Mouse anti-Canine, Human, FITC, Clone: H5C6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244247F

Catalog No. NBP244247F


Add to cart

Description

Description

CD133 Polyclonal antibody specifically detects CD133 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

CD133
H5C6
FITC
PBS with 0.05% Sodium Azide
AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1
Rabbit
IgG
0.1 ml
Autophagy, Cancer, Cardiovascular Biology, Cellular Signaling, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers
Primary
8842
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence
1.0 mg/mL
Flow Cytometry, Immunocytochemistry/Immunofluorescence
Purified
PROM1
A synthetic peptide made to an N-terminal portion of the human CD133 protein (between residues 1-100) [UniProt# O43490]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Immunogen has 100% homology to all isoforms of human CD133.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only