Learn More
CD63 Mouse anti-Rat, Alexa Fluor™ 700, Clone: AD1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP268077AF700100UL
Catalog No. N26807A7010
Add to cart
Description
Description
CD63 Monoclonal antibody specifically detects CD63 in Rat samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
CD63
AD1
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1lysosome-associated membrane glycoprotein 3, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, TSPAN30granulophysin
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Rat
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 700
50 mM sodium borate with 0.05% sodium azide
CD63
RBL-2H3 rat basophilic leukemia cells.
100 μL
Primary
967