Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD63 Mouse anti-Rat, PE, Clone: AD1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP268077PE100UL

Catalog No. N268077P10U


Add to cart

Description

Description

CD63 Monoclonal antibody specifically detects CD63 in Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD63
AD1
Flow Cytometry
CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1lysosome-associated membrane glycoprotein 3, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, TSPAN30granulophysin
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Rat
Flow Cytometry
PE
PBS with 0.05% sodium azide
CD63
RBL-2H3 rat basophilic leukemia cells.
100 μL
Primary
967
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit