Learn More
CD63 Rat anti-Human, Mouse, Alexa Fluor™ 750, Clone: 7G4.2E8, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP236567AF750
Catalog No. NP23667A70
Add to cart
Description
Description
CD63 Monoclonal antibody specifically detects CD63 in Human,Mouse samples. It is validated for Western Blot,Flow Cytometry,Flow Cytometry.
Specifications
Specifications
CD63
7G4.2E8
Western Blot, Flow Cytometry, Flow (Intracellular)
CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1lysosome-associated membrane glycoprotein 3, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, TSPAN30granulophysin
Rat
IgG2a Lambda
Autophagy, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis
Primary
967
Western Blot, Flow Cytometry
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
CD63
Partial recombinant mouse CD63 protein (between amino acids 100-250) [UniProt P41731]
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Mouse