Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD64 Mouse anti-Human, PE, Clone: 10.1, Invitrogen™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen CD6404

 View more versions of this product

Catalog No. 50-112-1255


Add to cart

Description

Description

Allophycocyanin (APC) is a stable and highly soluble phycobiliprotein that provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing exceptional quantum yields and molar extinction coefficients.

CD64 (FCGR1A) encodes a protein that plays an important role in the immune response. CD64 is a high-affinity Fc-gamma receptor and is one of three related gene family members located on chromosome 1. CD64 plays an important role in the immune response and its dysfunction has been studied in cervical adenitis and tetrasomy 21.
Specifications

Specifications

CD64
Monoclonal
PE
FCGR1A
Liquid
FCGR1A
Human CD64
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
10.1
PBS with BSA, sucrose and 0.1% sodium azide
P12314
RP11-196G18.2, CD64, CD64A, FCRI, FLJ18345, IGFR1, Fc fragment of IgG, high affinity Ia, receptor for CD64, Fc gamma receptor, Fc-gamma RI, Fc-gamma receptor 1
Mouse
IgG1
0.5 mL
4° C, store in dark
Primary
2209
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit