Learn More
CD65s Mouse anti-Human, PE, Clone: VIM2, BD Pharmingen™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565459
Catalog No. BDB565459
Add to cart
Description
Description
CD65s Monoclonal antibody specifically detects CD65s in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
CD65s
VIM2
Vol. Per test: 5μL
Purified
Murine
Mouse IgM, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions
Monoclonal
Flow Cytometry
PE
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Sialylated-CD65; Sialylated poly-N-acetyllactosamine; VIM2; VIM-2; M2
Human THP-1 Cell Line
50 Tests
Cell Biology
Primary
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.