Learn More
CD66 Mouse anti-Human, APC, Clone: CD66a-B1.1, eBioscience
Mouse Monoclonal Antibody
Manufacturer: Invitrogen 17066841
Description
Description: The CD66a-B1.1 monoclonal antibody reacts with human CD66a, c, d, and e, which are also known as CEACAM1, CEACAM6, CEACAM3 and CEA, respectively. CD66 is expressed on endothelial and epithelial cells, as well as on neutrophils and monocytes. Myeloma and cancer cells also express CD66. Moreover, expression of this receptor can be induced on T cells, B cells, and CD16-negative NK cells. CD66 exists as multiple splice variants that differ in the length of their cytoplasmic domain, which contains an immunoreceptor tyrosine-based inhibitory motif (ITIM) that mediates the inhibitory functions of this cell surface protein on T and NK cells. CD66 is involved in cell adhesion and growth, neutrophil activation, and signaling. Finally, studies have shown that the CD66 variants interact with each other homo- and heterotypically. Applications Reported: This CD66a-B1.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CD66a-B1.1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.06 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 µm post-manufacturing filtered.
The CD66 molecules are 180-200 kDa glycoproteins of carcinoembryonic antigen family. They are present on all blood granulocytes and some tissue macrophages, but are absent from other hematopoietic cells. The expression of CD66 increases significantly on granulocytes upon their activation. CD66a (BGP1, CEACAM1) and CD66d (CGM1, CEACAM3), as well as CEACAM2 and 4, are transmembrane proteins, whereas CD66c (CEAL, NCA, CEACAM6) and CD66e (CEA, CEACAM5), as well as CEACAM7 and 8, are anchored to the plasma membrane by C-terminal glycosylphosphatidylinositol (GPI) lipid moiety.Specifications
CD66 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
O75871, P06731, P40199, P13688, P40198 | |
pan CEACAM, Carcinoembryonic antigen-related cell adhesion molecule 1, BGP, BGP-1, CD66a, Biliary glycoprotein 1, Carcinoembryonic antigen-related cell adhesion molecule 3, CD66d, Carcinoembryonic antigen CGM1, CGM1, Carcinoembryonic antigen-related cell adhesion molecule 5, CEA, CD66e, Carcinoembryonic antigen-related cell adhesion molecule 6, CD66c, NCA, CEACAM-1, CEACAM-3, CEACAM-5, CEACAM-6 | |
Mouse | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Flow Cytometry | |
CD66a-B1.1 | |
APC | |
CEACAM5 | |
Liquid | |
CEACAM1, CEACAM3, CEACAM4, CEACAM5, CEACAM6 | |
IgG2a, kappa | |
25 Tests | |
4° C, store in dark, DO NOT FREEZE! | |
Primary | |
1048, 1084, 1089, 4680, 634 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.