Learn More
CD68 Mouse anti-Human, Clone: OTI4G1, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF802952
Catalog No. 50-166-6257
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CD68 (Macrosialin) is a 110 kDa integral membrane glycoprotein predomitly expressed on the intracellular lysomsomes of monocytes and macrophages and to a lesser extent by dendritic cells and peripheral blood granulocytes. Also, CD68 could play a role in phagocytic activities of tissue macrophages, both in intracellular lysosomal metabolism and extracellular cell-cell and cell-pathogen interactions. CD68 is expressed by interdigitating reticulum cells in tonsil and some histiocytic lymphoma or histiocytosis, acute myeloid leukemia (AML), and granulocytic sarcoma. Elevated expression of CD68 has been demonstrated on CD34+ cells in various human maligcies, including several Acute Myeloid Leukemia studies.
Specifications
Specifications
CD68
Monoclonal
Unconjugated
CD68
Lyophilized
CD68
Human recombit protein fragment corresponding to amino acids 22-319 of human CD68 produced in SF9 cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI4G1
PBS with 8% trehalose and no preservative; pH 7.3
P34810
GP110, LAMP4, SCARD1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
968