Learn More
CD68 Monoclonal Antibody (FA-11), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Supplier: Invitrogen 25068180
Description
Description: This FA-11 monoclonal antibody reacts with mouse CD68, known also as macrosialin. CD68 is a heavily O- and N- glycosylated type I integral membrane protein belonging to the sialomucin family and is closely related to the Lysosomal/endosomal-Associated Membrane Glycoproteins (LAMP). Although commonly used as a pan-macrophage marker, low levels of expression have also been observed in Langerhans cells and conventional dendritic cells. CD68 is mostly localized in the late endosomes and lysosomes with a smaller fraction circulating to the cell surface. In the steady state, only about 10-15% of CD68 is present at the surface. Cell stimulation increases expression of CD68, its transport to cell surface, and results in glycoform remodeling. CD68 binds oxidized LDL and is involved in the process of phagocytosis. The FA-11 antibody is not recommended for use in immunohistochemistry of formalin-fixed paraffin embedded tissues. Applications Reported: This FA-11 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This FA-11 antibody has been tested by intracellular staining and flow cytometric analysis of mouse bone marrow-derived macrophages using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online.
CD68 Monoclonal antibody specifically detects CD68 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD68 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P31996 | |
Cd68 | |
Affinity chromatography | |
RUO | |
12514 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
FA-11 | |
PE-Cyanine7 | |
Cd68 | |
CD68; CD68 antigen; CD68 molecule; GP110; Lamp4; macrophage antigen CD68; macrosialin; Scard1; scavenger receptor class D, member 1; similar to CD68 antigen | |
Rat | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.