Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD68/SR-D1 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI1E6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270381AF700

Catalog No. N270381A700


Add to cart

Description

Description

CD68/SR-D1 Monoclonal antibody specifically detects CD68/SR-D1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CD68/SR-D1
OTI1E6
Western Blot
Affinity Purified, Conjugated
CD68
Recombinant protein fragment corresponding to amino acids 22-319 of human CD68/SR-D1 (NP_001242)produced in SF9 cell.
Immunogen affinity purified
Cell Biology, Cytokine Research, Immunology
Primary
968.0
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit