Learn More
CD68/SR-D1 Mouse anti-Human, DyLight 350, Clone: 3F7D3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237265UV
Catalog No. NBP237265UV
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD68/SR-D1
3F7D3
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
CD68
Purified recombinant fragment of human CD68 (AA: 42-155) expressed in E. Coli.
Protein A or G purified
Cell Biology, Cytokine Research, Immunology
Primary
968
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human