Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD68/SR-D1 Mouse anti-Human, DyLight 350, Clone: OTI1E6, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270381UV
Description
CD68/SR-D1 Monoclonal antibody specifically detects CD68/SR-D1 in Human samples. It is validated for Western Blot.Specifications
CD68/SR-D1 | |
OTI1E6 | |
Western Blot | |
Affinity Purified, Conjugated | |
CD68 | |
Recombinant protein fragment corresponding to amino acids 22-319 of human CD68/SR-D1 (NP_001242)produced in SF9 cell. | |
Immunogen affinity purified | |
Cell Biology, Cytokine Research, Immunology | |
Primary | |
968.0 |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1 | |
Mouse | |
IgG2a | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title