Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD68/SR-D1 Mouse anti-Human, FITC, Clone: 3F7D3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237265F

Catalog No. NBP237265F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CD68/SR-D1
3F7D3
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
CD68
Purified recombinant fragment of human CD68 (AA: 42-155) expressed in E. Coli.
Protein A or G purified
Cell Biology, Cytokine Research, Immunology
Primary
968
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
FITC
PBS with 0.05% Sodium Azide
CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit