Learn More
CD68/SR-D1 Rat anti-Mouse, Alexa Fluor™ 532, Clone: FA-11, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233337AF532
Catalog No. NP233337A32
Add to cart
Description
Description
CD68/SR-D1 Monoclonal antibody specifically detects CD68/SR-D1 in Mouse samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications
Specifications
CD68/SR-D1
FA-11
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular)
CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1
Rat
IgG2a
Store at 4°C in the dark.
Monoclonal
Mouse
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CD68
Purified ConA acceptor glycoproteins from the P815 cell line.
0.1 mL
Primary
968