Learn More
CD69 Armenian Hamster anti-Mouse, BB700, Clone: H1.2F3, BD Horizon™
Armenian Hamster Monoclonal Antibody
Manufacturer:  BD Biosciences 566501
Catalog No. BDB566501
Add to cart
Description
Description
CD69 Monoclonal antibody specifically detects CD69 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD69
H1.2F3
BB700
Armenian Hamster
25 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG1, λ3
RUO
Primary
Mouse