Learn More
CD69 Mouse anti-Human, Alexa Fluor™ 350, Clone: 15B5G2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP225236AF350
Catalog No. NP22236A30
Add to cart
Description
Description
CD69 Monoclonal antibody specifically detects CD69 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
CD69
15B5G2
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3
Mouse
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CD69
Amino acids 103-199 of human CD69
0.1 mL
Primary
969