Learn More
CD69 Mouse anti-Human, PE-Cyanine5.5, Clone: CH/4, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD6918
Catalog No. 50-112-1456
Add to cart
Description
Description
The R-phycoerythrin (PE)-Cy5.5™ tandem conjugate permits simultaneous multicolor labeling and detection of multiple targets with excitation by a single excitation source-the 488 nm line of the argon-ion laser.
CD69 (AIM, Active Inducer Molecule) is a gp28/34 disulfide bonded homodimer with a molecular weight of 60 kDa under non-reducing conditions. CD69 contains one or two N linked oligosacaride and the molecule is present on activated platelets. In normal peripheral blood a variable percentage of cells express the CD69 antigen, and it is involved in lymphocyte signal transduction. Expression CD69 is induced upon activation of T lymphocytes, and may play a role in proliferation. Furthermore, the protein may act to transmit signals in natural killer cells and platelets. Alternative splicing results in multiple transcript variants of CD69. Diseases associated with CD69 dysfunction include coccidiodomycosis and asthma.
Specifications
Specifications
CD69
Monoclonal
PE-Cyanine5.5
CD69
Liquid
CD69
Human CD69
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
CH/4
PBS with BSA, sucrose and 0.1% sodium azide
Q07108
AIM, BL-AC/P26, CLEC2C, EA1, GP32/28, MLR-3, C-type lectin domain family 2, member C, CD69 antigen
Mouse
IgG2a
0.5 mL
4° C, store in dark
Primary
969