Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD69 Mouse anti-Human, Unconjugated, Clone: FN61, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen CD69
Applications Flow Cytometry, Immunofluorescence
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF22101MI
View Documents
MilliporeSigma
MABF2210100UG
100 μg
Each for $372.00
Add to cart
 
MABF22102MI
View Documents
MilliporeSigma
MABF221025UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

CD69 Monoclonal antibody specifically detects CD69 in Human samples. It is validated for Combined Interference Assay, Flow Cytometry, Inhibits Activity/Function, Immunofluorescence.
Specifications

Specifications

CD69
Unconjugated
Mouse
Early activation antigen CD69;Activation inducer molecule;AIM;BL-AC/P26;C-type lectin domain family 2 member C;EA1;Early T-cell activation antigen p60;GP32/28;Leukocyte surface antigen Leu-23;MLR-3
Activated B cells from human peripheral blood.
Primary
Stable for 1 year at -20°C from date of receipt.
Flow Cytometry, Immunofluorescence
Purified
Human
CD69, CLEC2C
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit