Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD69 Rabbit anti-Human, Alexa Fluor 647, Clone: 74, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289822AF647

Catalog No. NB109566

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CD69 Monoclonal antibody specifically detects CD69 in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

CD69
74
50 mM Sodium Borate
Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD69 (NP_001772.1; Ser 62-Lys 199).
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
Western Blot, ELISA
Alexa Fluor 647
Purified
Rabbit
IgG
0.1 mL
Innate Immunity
Primary
969
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit