Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD7 Monoclonal Antibody (eBio124-1D1 (124-1D1)), NovaFluor™ Red 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Specifications
Antigen | CD7 |
---|---|
Clone | eBio124-1D1 (124-1D1) |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H018T02R03
![]() |
LIFE TECHNOLOGIES
H018T02R03 |
25 Tests | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
H018T03R03
![]() |
LIFE TECHNOLOGIES
H018T03R03 |
100 Tests | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Flow CytometrySpecifications
CD7 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Cd7; CD7 antigen; CD7 antigen (p41); Cd7 molecule; GP40; LEU-9; p41 protein; T-cell antigen CD7; T-cell leukemia antigen; T-cell surface antigen Leu-9; Tp40; TP41 | |
CD7 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD7 |
eBio124-1D1 (124-1D1) | |
Flow Cytometry | |
NovaFluor Red 700 | |
Mouse | |
Human | |
P09564 | |
924 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title