Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD7 Monoclonal Antibody (eBio124-1D1 (124-1D1)), NovaFluor™ Yellow 610, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Specifications

Antigen CD7
Clone eBio124-1D1 (124-1D1)
Concentration 5 μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
H018T02Y03
Encompass
View Documents
LIFE TECHNOLOGIES
H018T02Y03
25 Tests This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
H018T03Y03
Encompass
View Documents
LIFE TECHNOLOGIES
H018T03Y03
100 Tests This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD7
5 μL/Test
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
Cd7; CD7 antigen; CD7 antigen (p41); Cd7 molecule; GP40; LEU-9; p41 protein; T-cell antigen CD7; T-cell leukemia antigen; T-cell surface antigen Leu-9; Tp40; TP41
CD7
Primary
4° C, store in dark, DO NOT FREEZE!
CD7
eBio124-1D1 (124-1D1)
Flow Cytometry
NovaFluor Yellow 610
Mouse
Human
P09564
924
IgG1 κ
Affinity chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit