Learn More
CD7 Mouse anti-Human, PE, Clone: CD7-6B7, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD0704
Catalog No. 50-112-1198
Add to cart
Description
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
CD7 (gp40, Leu9) is a 40 kDa member of the immunoglobulin gene superfamily. CD7 contains N-terminal amino acids 1-107 are highly homologous to Ig kappa-L chains whereas the carboxy-terminal region of the extracellular domain is proline-rich and has been postulated to form a stalk from which the Ig domain projects. CD7 is expressed on the majority of immature and mature T-lymphocytes, and T cell leukemia. Further, CD7 is also found on natural killer cells, a small subpopulation of normal B cells and on maligt B cells. Cross-linking surface CD7 positively modulates T cell and NK cell activity as measured by calcium fluxes, expression of adhesion molecules, cytokine secretion and proliferation. CD7 associates directly with phosphoinositol 3 ft.-kinase. CD7 ligation induces production of D-3 phosphoinositides and tyrosine phosphorylation. Expression of CD7 is an important marker used in leukemia diagnostics.
Specifications
Specifications
CD7
Monoclonal
PE
CD7
Liquid
CD7
Human CD7
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
CD7-6B7
PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide
P09564
GP40, LEU-9, TP41, Tp40
Mouse
IgG2a
0.5 mL
4° C, store in dark
Primary
924