Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD7 Rabbit anti-Human, Alexa Fluor 488, Clone: 307, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289749AF488

Catalog No. NB109120


Add to cart

Description

Description

CD7 Monoclonal specifically detects CD7 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CD7
307
50 mM Sodium Borate
CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD7 (NP_006128.1; Met1-Pro180).
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 488
Purified
Rabbit
IgG
0.1 mL
Cytokine Research, Signal Transduction
Primary
924
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit