Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD7, Rabbit anti-Human, Clone: 3A1E, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP2810500.025MG
Description
CD7 Monoclonal specifically detects CD7 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional.Specifications
CD7 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry, Functional | |
CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Recognises human CD7. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG κ |
Flow Cytometry, Functional Assay | |
3A1E | |
Unconjugated | |
PBS | |
CD7 | |
Recombinant monoclonal antibody to CD7. | |
0.025 mg | |
Cytokine Research, Signal Transduction | |
924 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction