Learn More
CD70 Monoclonal Antibody (FR70), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$416.00
Specifications
Antigen | CD70 |
---|---|
Clone | FR70 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-159-80
|
Invitrogen
46070182 |
100 μg |
Each of 1 for $416.00
|
|
|||||
Description
Description: The FR70 monoclonal antibody reacts with mouse CD70, a 30-33 kDa type II transmembrane glycoprotein. CD70 is a member of the TNF family and is expressed by activated mouse T and B lymphocytes. Interaction of CD70 with CD27, its ligand on T cells, is important in T-B cell crosstalk and plays a role in costimulation and immune regulation. Applications Reported: This FR70 antibody has been reported for use in flow cytometric analysis. Applications Tested: This FR70 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
CD70 Monoclonal antibody specifically detects CD70 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD70 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
O55237 | |
Cd70 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Cd70 |
FR70 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Rat | |
Mouse | |
21948 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.