Learn More
CD71 (Transferrin Receptor) Mouse anti-Human, APC, Clone: OKT9 (OKT-9), eBioscience™
Mouse Monoclonal Antibody
Manufacturer: Invitrogen 17071941
Description
Description: The OKT9 monoclonal antibody reacts with human CD71, a 170-180 kDa type II transmembrane protein. CD71, the transferrin receptor, exists as a homodimer on the cell surface and is essential for cellular growth. CD71 is expressed by immature proliferating cells and at low levels on resting mature lymphocytes. Antigen or mitogen stimulation of T and B cells upregulates the expression of CD71. Applications Reported: This OKT9 (OKT-9) antibody has been reported for use in flow cytometric analysis. Applications Tested: This OKT9 (OKT-9) antibody has been pre-titrated and tested by flow cytometric analysis of stimulated human peripheral blood cells. This can be used at 5 µL (0.06 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system (By similarity). Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregcy, irritable bowel syndrome and in HIV patients.Specifications
CD71 (Transferrin Receptor) | |
Monoclonal | |
5 μL/Test | |
Liquid | |
TFRC | |
CD71, T9, TFR, TFR1, TR, TRFR, p90, transferrin receptor protein 1 | |
TFRC | |
IgG1, kappa | |
Primary | |
25 Tests | |
4° C, store in dark, DO NOT FREEZE! | |
Antibody |
Flow Cytometry | |
OKT9 (OKT-9) | |
APC | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P02786 | |
7037 | |
Mouse | |
Monoclonal | |
Affinity chromatography | |
RUO | |
Human |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.