Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD72 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093559B
Description
CD72 Polyclonal antibody specifically detects CD72 in Human samples. It is validated for Western BlotSpecifications
CD72 | |
Polyclonal | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
Biotin | |
CD72 antigenB-cell differentiation antigen CD72, CD72 molecule, CD72b, Lyb-2, LYB2lyb-2 | |
Synthetic peptide corresponding to residues 25-37 [LGQDPGADDDGEI] of the 42 kDa human CD72 protein. | |
0.1 mL | |
Adaptive Immunity, Immunology | |
971 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction