Learn More
CD72 Mouse anti-Human, eFluor® 660, Clone: J4-117, eBioscience™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen 50072942
Catalog No. 50-112-4284
Add to cart
Description
Description
Description: The monoclonal antibody J4-117 reacts with human CD72, a 45 kDa type II transmembrane glycoprotein that belongs to the C-type lectin superfamily. This glycoprotein is expressed on B cells at all stages of maturation except for plasma cells. CD72 is also expressed on dendritic cells, mast cells, and some macrophages. This receptor contains an immune-receptor tyrosine-base inhibition motif (ITIM) that associates with a tyrosine phosphatase, SHP-1. CD72 negatively regulates the major signal transduction pathways activated by the B cell receptor (BCR). Studies have demonstrated that the ligand for CD72 is CD100 (also known as Semaphorin 4D). CD72 can also react with CD5 to provide signaling between T and B cells. Finally, this receptor also prevents naive B cells from producing Ig and differentiating into B cells, and plays a role in controlling autoimmunity by enhancing apoptosis after Ag-specific BCR stimulation. Applications Reported: This J4-117 antibody has been reported for use in flow cytometric analysis. Applications Tested: This J4-117 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.06 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 µm post-manufacturing filtered. CD72 is a transmembrane glycoprotein expressed as a homodimer especially in B cells, but also in other antigen presenting cells such as dendritic cells and macrophages. Through one of its immunoreceptor tyrosine-based inhibitory motives (ITIMs), CD72 interacts with tyrosine phosphatase SHP-1, thereby suppressing B cell responsiveness. Binding of CD72 with its ligand CD100 (Sema4D) prevents BCR association and phosphorylation of CD72 and results in dissociation of SHP-1 from CD72, thus enables B cell activation.
Specifications
Specifications
CD72
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P21854
RP11-331F9.9, CD72b, LYB2
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
J4-117
eFluor™ 660
CD72
Liquid
CD72
IgG2b, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
971
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only.