Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD72 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093559SS
Description
CD72 Polyclonal specifically detects CD72 in Human samples. It is validated for Western Blot.Specifications
CD72 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
CD72 antigenB-cell differentiation antigen CD72, CD72 molecule, CD72b, Lyb-2, LYB2lyb-2 | |
Rabbit | |
40.2 kDa | |
0.025 mg | |
Adaptive Immunity, Immunology | |
971 | |
Human | |
IgG |
Western Blot | |
1 mg/mL | |
Western Blot 1:100-1:2000 | |
P21854 | |
CD72 | |
Synthetic peptide corresponding to residues 25-37 [LGQDPGADDDGEI] of the 42 kDa human CD72 protein. | |
Affinity Purified | |
RUO | |
Primary | |
CD72 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction