Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD72 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$177.16 - $378.00
Specifications
Antigen | CD72 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10093559
|
Novus Biologicals
NB10093559 |
0.1 mg |
Each for $378.00
|
|
NB017912
|
Novus Biologicals
NB10093559SS |
0.025 mg |
Each for $177.16
|
|
Description
CD72 Polyclonal specifically detects CD72 in Human samples. It is validated for Western Blot.Specifications
CD72 | |
Western Blot 1:100-1:2000 | |
Polyclonal | |
Rabbit | |
Adaptive Immunity, Immunology | |
PBS with 0.02% Sodium Azide | |
CD72 antigenB-cell differentiation antigen CD72, CD72 molecule, CD72b, Lyb-2, LYB2lyb-2 | |
CD72 | |
IgG | |
Affinity Purified | |
CD72 |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
P21854 | |
971 | |
Synthetic peptide corresponding to residues 25-37 [LGQDPGADDDGEI] of the 42 kDa human CD72 protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
40.2 kDa |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title