Learn More
CD73 Mouse anti-Human, NovaFluor™ Red 700, Clone: AD2, Invitrogen™
Mouse Monoclonal Antibody
Specifications
Antigen | CD73 |
---|---|
Clone | AD2 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H016T02R03
![]() |
LIFE TECHNOLOGIES
H016T02R03 |
25 Tests | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
H016T03R03
![]() |
LIFE TECHNOLOGIES
H016T03R03 |
100 Tests | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
This AD2 monoclonal antibody reacts with human CD73, a 5'-ectonucleotidase that converts 5'-adenosine monophosphate to adenosine. CD73 is expressed on the surface of endothelial cells, as well as B and T cells, including some CD4+Foxp3+ regulatory T cells. Adenosine production by these cells has been linked to the inhibition of CD4 T cell effector functions such as proliferation and cytokine secretion. Applications Reported: This AD2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This AD2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.4 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Each NovaFluor conjugate or kit is shipped with CellBlox Blocking Buffer. Excitation: 639 nm; Emission: 700 nm; Laser: 633-640 nm (Red) Laser NovaFluor conjugates are based on Phiton™ technology utilizing novel nucleic acid dye structures that allow for engineered fluorescent signatures with consideration for spillover and spread impacts.
CD73 Monoclonal antibody specifically detects CD73 in Human samples. It is validated for Flow CytometrySpecifications
CD73 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
2210401F01Rik; 5 nucleotidase, ecto; 5' nucleotidase, ecto; 5'-NT; 5'-nucleotidase; 5'-nucleotidase ecto; 5'-nucleotidase, ecto (CD73); AI447961; CALJA; CD73; E5NT; ecto-5'-nucleotidase; eN; eNT; NT; Nt5; Nt5e; NTE; Purine 5-Prime-Nucleotidase | |
NT5E | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
NT5E |
AD2 | |
Flow Cytometry | |
NovaFluor Red 700 | |
Mouse | |
Human | |
P21589 | |
4907 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.