Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD74 Mouse anti-Human, Alexa Fluor 647, Clone: 1B8, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259696AF647
Description
CD74 Monoclonal specifically detects CD74 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CD74 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD74 | |
Human CD74 invariant chain synthetic peptide. | |
100 μL | |
Primary | |
Detects approx 220kDa in extracts of cells transiently transfected with TrpM7. No cross-reactivity against TrpM6. | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1B8 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, MHC HLA-DR gamma chain, p33 | |
Mouse | |
Protein G purified | |
Immunology | |
972 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title