Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD74 Mouse anti-Human, PE-Cyanine7, Clone: 3D7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP259697PECY7

Catalog No. N259697PEC7


Add to cart

Description

Description

CD74 Monoclonal antibody specifically detects CD74 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD74
3D7
Flow Cytometry
Conjugated, Purified
CD74
Human CD74 invariant chain synthetic peptide.
Protein G purified
Immunology
Primary
972.0
Human
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, MHC HLA-DR gamma chain, p33
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Detects approx 80, 55kDa. No cross-reactivity against Cav Beta4.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit