Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD75/ST6GAL1 Mouse anti-Human, Unconjugated, Clone: HH2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS193025UL
Description
CD75/ST6GAL1 Monoclonal antibody specifically detects CD75/ST6GAL1 in Human samples. It is validated for Flow Cytometry, Inhibition.Specifications
CD75/ST6GAL1 | |
Monoclonal | |
Unconjugated | |
Beta-galactoside alpha-2;6-sialyltransferase 1;EC:2.4.99.1;Alpha 2;6-ST 1;-cell antigen CD75;CMP-N-acetylneuraminate-beta-galactosamide-alpha-2;6-sialyltransferase 1;ST6Gal I;ST6GalI;Sialyltransferase 1 | |
Mouse | |
46.61 kDa calculated. | |
25 μL | |
Primary | |
Human | |
Purified |
Flow Cytometry, Inhibition Assays | |
HH2 | |
NP_003023 | |
ST6GAL1, SIAT1 | |
Lysate from B-cells derived from lymphoma patient. | |
Ligatrap | |
Signaling | |
Clone HH2 is a mouse monoclonal antibody that detects human beta-galactoside alpha-2,6-sialyltransferase 1 (CD75). | |
Stable for 1 year at -20°C from date of receipt. | |
IgM κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction