Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD75/ST6GAL1 Mouse anti-Human, Unconjugated, Clone: HH2, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen CD75/ST6GAL1
Applications Flow Cytometry, Inhibition Assays
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS19301MI
View Documents
MilliporeSigma
MABS1930100UL
100 μL
Each for $372.00
Add to cart
 
MABS19302MI
View Documents
MilliporeSigma
MABS193025UL
25 μL
Each for $147.00
Add to cart
 
Description

Description

CD75/ST6GAL1 Monoclonal antibody specifically detects CD75/ST6GAL1 in Human samples. It is validated for Flow Cytometry, Inhibition.
Specifications

Specifications

CD75/ST6GAL1
Unconjugated
Mouse
Beta-galactoside alpha-2;6-sialyltransferase 1;EC:2.4.99.1;Alpha 2;6-ST 1;-cell antigen CD75;CMP-N-acetylneuraminate-beta-galactosamide-alpha-2;6-sialyltransferase 1;ST6Gal I;ST6GalI;Sialyltransferase 1
Lysate from B-cells derived from lymphoma patient.
Primary
Stable for 1 year at -20°C from date of receipt.
Flow Cytometry, Inhibition Assays
Purified
Human
ST6GAL1, SIAT1
Monoclonal
Ligatrap
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit