Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79a Mouse anti-Mouse, Clone: 24C2.5, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 14079180

 View more versions of this product

Catalog No. 50-127-04


Add to cart

Description

Description

Description: This 24C2.5 monoclonal antibody reacts with the intracellular tail of mouse CD79a (also known as Ig alpha or mb-1), a signaling component of the B cell receptor (BCR). CD79a heterodimerizes with CD79b (Ig beta, B29), and together with surface Ig, make up the BCR. Both CD79 subunits consist of a single extracellular Ig domain, a transmembrane domain, and an intracellular signaling domain. CD79a is expressed almost exclusively on B cells, as well as in most B lineage acute lymphoblastic leukemias. The cytoplasmic domain of CD79a contains immunoreceptor tyrosine-based activation motifs (ITAMs), which constitute the signal transducing portions of the BCR. Tyrosine phosphorylation of these ITAMs by Syk and Lyn initiates numerous signaling cascades, resulting in B cell activation, proliferation, and differentiation. Applications Reported: This 24C2.5 antibody has been reported for use in immunoblotting (WB). Applications Tested: This 24C2.5 antibody has been tested by western blot of reduced mouse spleen lysates. This can be used at 1-10 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered.

CD79a is a disulphide-linked heterodimer that includes B29 (CD79b) polypeptide. CD79a is a B lymphocyte antigen receptor with an antigen-specific surface component Ig (immunoglobulin) that associates with Ig-alpha and Ig-beta, necessary elements for the expression and function of the B-cell antigen receptor. CD79a first appears at pre-B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines. Diseases associated with CD79a include Agammaglobulinemia 3 and Agammaglobulinemia, Non-Bruton type.
Specifications

Specifications

CD79a
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P11911
IGA, MB-1
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Immunohistochemistry, Western Blot
24C2.5
Unconjugated
Cd79a
Liquid
Cd79a
IgG1, kappa
25 μg
4° C
Primary
12518
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.