Learn More
CD79A Mouse anti-Human, Clone: IGA/1406, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB15073
Catalog No. 89-035-093
Add to cart
Description
Description
Mouse monoclonal antibody raised against synthetic peptide of human CD79A.
  • High specificity
  • High reactivity
Specifications
Specifications
CD79A
IGA/1406
Mouse monoclonal antibody raised against synthetic peptide of human CD79A.
In 10 mM PBS.
Liquid
CD79A
A synthetic peptide corresponding to 15 residues of human CD79A.
Protein A/G purification
RUO
Primary
973
Flow Cytometry, Immunofluorescence, Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
P11912
IGA|MB-1
Mouse
IgG3
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)