Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79A Rabbit anti-Human, Mouse, Rat, Allophycocyanin/Cy7, Clone: IGA/1688R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254469APCCY7

Catalog No. N254469APC7


Add to cart

Description

Description

CD79A Monoclonal antibody specifically detects CD79A in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD79A
IGA/1688R
Flow Cytometry
Conjugated, Purified
CD79A
A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Protein A purified
Adaptive Immunity, Cell Biology, Immunology, Phospho Specific
Primary
973.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
CD79a antigen, CD79A antigen (immunoglobulin-associated alpha), CD79a molecule, immunoglobulin-associated alpha, IGAB-cell antigen receptor complex-associated protein alpha chain, Ig-alpha, MB1, MB-1, MB-1 membrane glycoprotein, Membrane-bound immunoglobulin-associated protein, Surface IgM-associated protein
Rabbit
IgG Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit