Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79B Mouse anti-Human, Clone: IGB/1843, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30741810UG

 View more versions of this product

Catalog No. NB122309


Add to cart

Description

Description

CD79B Monoclonal antibody specifically detects CD79B in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

CD79B
IGB/1843
Unconjugated
10 mM PBS with 0.05% BSA
B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein
Recombinant human CD79B protein fragment (around aa29-159) (exact sequence is proprietary)
Protein A or G purified
RUO
Primary
974
Western Blot, ELISA
0.2 mg/ml
Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml
Purified
Mouse
IgG1 Kappa
100 μg
Store at 4°C.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit