Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79B Mouse anti-Human, Clone: IGB/1844, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen CD79B
Clone IGB/1844
Concentration 0.2 mg/ml
Dilution ELISA 2-4 ug/ml, Protein Array
Applications ELISA, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122312
View Documents
Novus Biologicals
NBP30741920UG
20 μg
Each for $138.37
Add to cart
 
NB122311
View Documents
Novus Biologicals
NBP30741910UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

CD79B Monoclonal antibody specifically detects CD79B in Human samples. It is validated for ELISA, Protein Array
Specifications

Specifications

CD79B
0.2 mg/ml
ELISA, Peptide Array
Purified
RUO
10 mM PBS with 0.05% BSA
974
IgG1 Kappa
Primary
Store at 4°C.
IGB/1844
ELISA 2-4 ug/ml, Protein Array
Unconjugated
Mouse
Human
B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein
Recombinant human CD79B protein fragment (around aa 29-159) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit