Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

CD79B Rabbit anti-Human, Clone: IGB/2940R, Novus Biologicals™

Rabbit Monoclonal Antibody

Specifications

Antigen CD79B
Clone IGB/2940R
Concentration 0.2 mg/ml
Dilution ELISA 2-4 ug/ml
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122353
View Documents
Novus Biologicals
NBP30744010UG
100 μg
Each for $406.52
N/A
NB122354
View Documents
Novus Biologicals
NBP30744020UG
20 μg
Each for $159.82
N/A
Description

Description

CD79B Monoclonal antibody specifically detects CD79B in Human samples. It is validated for ELISA
Specifications

Specifications

CD79B
0.2 mg/ml
ELISA
Purified
RUO
10 mM PBS with 0.05% BSA
974
IgG
Primary
Store at 4°C.
IGB/2940R
ELISA 2-4 ug/ml
Unconjugated
Rabbit
Human
B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein
Recombinant human CD79B protein fragment (around aa29-159) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit