Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79B Rabbit anti-Human, Unconjugated, Clone: IGB/3170R, Abnova™

Rabbit Monoclonal Antibody.

Manufacturer:  Abnova Corporation RAB00681

Catalog No. 89-339-720


Add to cart

Description

Description

CD79B Monoclonal antibody specifically detects CD79B in Human samples. It is validated for ELISA, IHC-P.
Specifications

Specifications

CD79B
IGB/3170R
CD79b molecule, immunoglobulin-associated beta
Liquid
Rabbit
IgG
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunohistochemistry (Paraffin)
Unconjugated
CD79B
B29, IGB
Recombinant protein corresponding to amino acids 29-159 of human CD79B
100 μg
Primary
974
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit