Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 alpha Rat anti-Mouse, APC, Clone: AP-MAB0708, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212183APC0.1ML

Catalog No. NB2183APC01

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CD8 Monoclonal antibody specifically detects CD8 in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD8 alpha
AP-MAB0708
Flow Cytometry
Purified
CD8A
This antibody was produced from a hybridoma (mouse myeloma) fused with spleen cells from a rat immunized with mouse Thymus/Spleen Tissues.
27 kDa
0.1 ml
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary
925
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
CD8, CD8 antigen, alpha polypeptide (p32), CD8a antigen, CD8a molecule, Leu2, Leu2 T-lymphocyte antigen, MAL, OKT8 T-cell antigen, p32, RPA-T8, RPA-T8 antibody flow, RPA-T8 CD8, RPA-T8 Clone, RPA-T8 Flow, T cell co-receptor, T8 T-cell antigen, T-cell antigen Leu2, T-cell surface glycoprotein CD8 alpha chain, T-lymphocyte differentiation antigen T8/Leu-2
Rat
IgG2b
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only