Learn More
CD8 alpha Rat anti-Mouse, DyLight 405, Clone: AP-MAB0708, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212183V0.1ML
Catalog No. NB212183V01
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD8 Monoclonal antibody specifically detects CD8 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
CD8 alpha
AP-MAB0708
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
CD8A
This antibody was produced from a hybridoma (mouse myeloma) fused with spleen cells from a rat immunized with mouse Thymus/Spleen Tissues.
27 kDa
0.1 ml
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary
925
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
CD8, CD8 antigen, alpha polypeptide (p32), CD8a antigen, CD8a molecule, Leu2, Leu2 T-lymphocyte antigen, MAL, OKT8 T-cell antigen, p32, RPA-T8, RPA-T8 antibody flow, RPA-T8 CD8, RPA-T8 Clone, RPA-T8 Flow, T cell co-receptor, T8 T-cell antigen, T-cell antigen Leu2, T-cell surface glycoprotein CD8 alpha chain, T-lymphocyte differentiation antigen T8/Leu-2
Rat
IgG2b
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Mouse