Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (53-6.7) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody has been used in 16 publications
Supplier: Novus Biologicals NBP149045
Description
CD8 Monoclonal specifically detects CD8 in Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Cell depletion, Inhibition of T Cell Function, CyTOF-ready.Specifications
CD8 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature, Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 24565643), Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Cell depletion, Inhibition of T Cell Function, CyTOF-ready | |
CD8A | |
Mouse thymus or spleen. | |
Affinity Purified | |
RUO | |
925 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
53-6.7 | |
Unconjugated | |
53-6.7, 53-6.7 CD8, 53-6.7 CD8 Alpha, 53-6.7 CD8 Aplha, 53-6.7 Clone, CD8, CD8a molecule, Cytotoxic T cell marker, Cytotoxic T lymphocyte marker, LEU2, p32, T cell marker | |
Rat | |
27 kDa | |
0.5 mg | |
Primary | |
Mouse, Rat | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction