Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CD8 Antibody Pair [HRP]

Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2796125PLATES

 View more versions of this product

Catalog No. NB010264


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Ferret CD8.
Specifications

Specifications

Sandwich ELISA
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Ferret CD8.
78 pg/mL
925
RUO
CD8
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers
Ferret
CD8A
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 5000 pg/mL is recommended.
HRP
Antibody Pairs
Sandwich ELISA
  • Standard: Each vial contains 110 ng of recombinant CD8.
  • Capture Antibody: 1 mg/mL of rabbit anti-CD8 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of mouse anti-CD8 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Storage is content dependent.
    Rabbit/Mouse
    5 Plates
    78-5000 pg/mL
    CD8, CD8a molecule, LEU2, p32
    Conjugated
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit