Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

CD8 beta Rat anti-Mouse, Clone: H35-17.2, Novus Biologicals™

Rat Monoclonal Antibody

Specifications

Antigen CD8 beta
Clone H35-17.2
Concentration 0.2 mg/ml
Dilution Flow Cytometry 0.5-1 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Functional
Applications Flow Cytometry, Immunofluorescence, Functional Assay
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122135
View Documents
Novus Biologicals
NBP30733110UG
100 μg
Each for $352.89
N/A
NB122136
View Documents
Novus Biologicals
NBP30733120UG
20 μg
Each for $138.37
N/A
Description

Description

CD8 beta Monoclonal antibody specifically detects CD8 beta in Mouse samples. It is validated for Flow Cytometry, Immunofluorescence, Functional Assay
Specifications

Specifications

CD8 beta
0.2 mg/ml
Flow Cytometry, Immunofluorescence, Functional Assay
Purified
RUO
Mouse
CD8 antigen, beta polypeptide 1 (p37), CD8b antigen, CD8b molecule, CD8B1P37, LEU2, LY3, LYT3, MGC119115, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain
Five day mixed leukocyte culture population, C57BL/6 anti-BALB/c
Monoclonal
Protein A or G purified
H35-17.2
Flow Cytometry 0.5-1 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Functional
Unconjugated
Rat
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers
10 mM PBS with 0.05% BSA
926
IgG2b Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit