Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Monoclonal Antibody (49-31.1), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA170076
Description
MA1-70076 detects CD8a in mouse samples. MA1-70076 has been successfully used in flow cytometry procedures. The MA1-70076 immunogen is 129/ReJ.
CD8 (Cluster of Differentiation 8) is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediate efficient cell-cell interactions within the immune system. The CD8 antigen acts as a co-receptor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. CD8 is found on a T cell subset of normal cytotoxic/suppressor cells which make up approximately 20-35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer (NK) cells, subpopulations of peripheral blood null cells, thymocytes and bone marrow cells. The CD8 co-receptor functions as either a homodimer composed of two alpha chains, or as a disulfide-linked heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. The majority of CD8+ T cells express CD8 as a alpha/beta heterodimer. In HIV, the HIV-2 envelope glycoprotein binds CD8 alpha chain (but not the beta chain).Specifications
CD8 | |
Monoclonal | |
1 mg/mL | |
Cd8a | |
CD8, Leu-2, MAL, p32, CD8a, CD8b, CD8alpha, CD8beta | |
Mouse | |
Protein G | |
RUO | |
12525 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry | |
49-31.1 | |
Unconjugated | |
P01731 | |
Cd8a | |
129/ReJ. | |
250 μg | |
Primary | |
Mouse | |
Antibody | |
IgG3 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title