Learn More
CD8 Mouse anti-Human, Alexa Fluor™ 594, Clone: 37006R, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB1509RT100UG
Catalog No. FB1509RT100
Add to cart
This item is not returnable. View return policy
Description
Description
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD8
37006R
Flow Cytometry 0.25-1 μg/106 cells
NP_741969
CD8, CD8A, CD8a molecule, LEU2, p32
Mouse myeloma cell line NS0-derived recombinant human CD8 alpha , Ser22-Val198, Accession # NP_741969
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human CD8 alpha in direct ELISAs.
Flow Cytometry
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG2b
100 μg
Primary
925
Human