Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Mouse anti-Human, Baboon, Cynomolgus, Rhesus, BUV563, Clone: RPA-T8, BD Horizon™
Mouse Monoclonal Antibody
$240.00 - $456.00
Specifications
Antigen | CD8 |
---|---|
Clone | RPA-T8 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 563 |
Description
CD8 Monoclonal antibody specifically detects CD8 in Human,Baboon,Cynomolgus,Rhesus samples. It is validated for Flow Cytometry.Specifications
CD8 | |
Flow Cytometry | |
Brilliant Ultraviolet 563 | |
RUO | |
IgG1 κ | |
Refrigerate (2 to 8°C) |
RPA-T8 | |
Monoclonal | |
Mouse | |
Human, Baboon, Cynomolgus, Rhesus Monkey | |
Primary | |
CD8 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title